ContactUsLinkComponent
Interferon-alpha 1a Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 19.4 kDa.The Interferon-alpha 1a contains Valine residue at position 114.The 1 * 20 µG
Catalog # BIRBORB80385-20
Supplier:  Biorbyt
Interferon-alpha 1a Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 19.4 kDa.The Interferon-alpha 1a contains Valine residue at position 114.The 1 * 20 µG
Catalog # BIRBORB80385-20
Supplier:  Biorbyt