ContactUsLinkComponent
Prolactin Receptor Ovine Extra Celleular Domain Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 213 amino acids and having a molecular mass of 24.4 kDa. The Prolactin Receptor is purified by proprietary chro 1 * 10 µG
Catalog # BIRBORB80567-10
Supplier:  Biorbyt
Prolactin Receptor Ovine Extra Celleular Domain Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 213 amino acids and having a molecular mass of 24.4 kDa. The Prolactin Receptor is purified by proprietary chro 1 * 10 µG
Catalog # BIRBORB80567-10
Supplier:  Biorbyt