ContactUsLinkComponent
GMCSF Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 127 amino acids fragment (18-144) a nd having a molecular mass of 18.98kDa with an amino-terminal hexahistidine tag. GMCSF Human Recombinant His 1 * 5 µG
Katalogové číslo BIRBORB80374-5
Supplier:  Biorbyt
GMCSF Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 127 amino acids fragment (18-144) a nd having a molecular mass of 18.98kDa with an amino-terminal hexahistidine tag. GMCSF Human Recombinant His 1 * 5 µG
Katalogové číslo BIRBORB80374-5
Supplier:  Biorbyt