ContactUsLinkComponent
Placenta Growth Factor-2 Human Recombinant produced in insect cells is homodimer, glycosylated polypeptide chain containing x 152 am ino acids and having total molecular mass of 44 kDa. The PLGF-2 is purified by proprietary chromatographic t 1 * 2 µG
Catalog # BIRBORB80557-2
Supplier:  Biorbyt
Placenta Growth Factor-2 Human Recombinant produced in insect cells is homodimer, glycosylated polypeptide chain containing x 152 am ino acids and having total molecular mass of 44 kDa. The PLGF-2 is purified by proprietary chromatographic t 1 * 2 µG
Catalog # BIRBORB80557-2
Supplier:  Biorbyt