ContactUsLinkComponent
Placental Lactogen Caprine Recombinant, is a single polypeptide chain containing 199 amino acids and an additional Ala at the N-term inus having a molecular mass of 23 kDa. Placental Lactogen Recombinant Goat is purified by proprietary chrom 1 * 50 µG
Catalog # BIRBORB80555-50
Supplier:  Biorbyt
Placental Lactogen Caprine Recombinant, is a single polypeptide chain containing 199 amino acids and an additional Ala at the N-term inus having a molecular mass of 23 kDa. Placental Lactogen Recombinant Goat is purified by proprietary chrom 1 * 50 µG
Catalog # BIRBORB80555-50
Supplier:  Biorbyt