ContactUsLinkComponent
Placental Lactogen Ovine Recombinant, is a single polypeptide chain containing 199 amino acids and an additional Ala at the N-termin us having a molecular mass of 23 kDa. Placental Lactogen Recombinant is purified by proprietary chromatograp 1 * 50 µG
Catalog # BIRBORB80554-50
Supplier:  Biorbyt
Placental Lactogen Ovine Recombinant, is a single polypeptide chain containing 199 amino acids and an additional Ala at the N-termin us having a molecular mass of 23 kDa. Placental Lactogen Recombinant is purified by proprietary chromatograp 1 * 50 µG
Catalog # BIRBORB80554-50
Supplier:  Biorbyt