AURKB Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 364 amino acids (1-344) and h aving a molecular mass of 41.4kDa. AURKB is fused to 20 a.a. His-Tag at N-terminus and purified by proprieta 1 * 2 µG
Catalog # BIRBORB80775-2
Supplier: Biorbyt
Frequently Bought Together