ContactUsLinkComponent
PDGF-AA Human Recombinant produced in Yeast is homodimeric, glycosilated, polypeptide chain containing x 110 amino acids and having total molecular mass of 34 kDa. PDGF-AA is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80539-10
Supplier:  Biorbyt
PDGF-AA Human Recombinant produced in Yeast is homodimeric, glycosilated, polypeptide chain containing x 110 amino acids and having total molecular mass of 34 kDa. PDGF-AA is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80539-10
Supplier:  Biorbyt