ContactUsLinkComponent
Omentin Human Recombinant is produced in E.coli by recombinant DNA technology is a single, polypeptide chain containing 294 amino ac ids and having a molecular mass of 32.7 kDa. Recombinant Human Omentin contains His tag fused at N-Terminus. 1 * 10 µG
  BIRBORB80534-10
 :  Biorbyt
Omentin Human Recombinant is produced in E.coli by recombinant DNA technology is a single, polypeptide chain containing 294 amino ac ids and having a molecular mass of 32.7 kDa. Recombinant Human Omentin contains His tag fused at N-Terminus. 1 * 10 µG
  BIRBORB80534-10
 :  Biorbyt
Frequently Bought Together