ContactUsLinkComponent
LGALS9 produced in E.coli is a single, non-glycosylated polypeptide chain containing 168 amino acids (1-148 a.a.) and having a molec ular mass of 18.5 kDa. Galectin 9 is fused to 20 amino acid His-Tag at N-terminus and purified by proprietar 1 * 10 µG
Catalog # BIRBORB80335-10
Supplier:  Biorbyt
LGALS9 produced in E.coli is a single, non-glycosylated polypeptide chain containing 168 amino acids (1-148 a.a.) and having a molec ular mass of 18.5 kDa. Galectin 9 is fused to 20 amino acid His-Tag at N-terminus and purified by proprietar 1 * 10 µG
Catalog # BIRBORB80335-10
Supplier:  Biorbyt