AK2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 259 amino acids and having a mo lecular mass of 28.6 kDa. AK2 is fused to 20 a.a. His-Tag at N-terminus and purified by proprietary chromato 1 * 10 µG
Catalog # BIRBORB80770-10
Supplier: Biorbyt
Frequently Bought Together