ContactUsLinkComponent
Thymopentin has molecular formula of C30H49N9O9, H-Arg-Lys-Asp-Val-Tyr-OH having Mw of 679.8 Dalton. 1 * 100 mg
Catalog # BIRBORB80763-100
Supplier:  Biorbyt
Thymopentin has molecular formula of C30H49N9O9, H-Arg-Lys-Asp-Val-Tyr-OH having Mw of 679.8 Dalton. 1 * 100 mg
Catalog # BIRBORB80763-100
Supplier:  Biorbyt