ContactUsLinkComponent
LGALS2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 152 amino acids (1-132 a.a.) and having a molecular mass of 16.8 kDa. The LGALS2 is fused to 20 amino acid His-Tag at N-terminus and pur 1 * 5 µG
Catalog # BIRBORB80329-5
Supplier:  Biorbyt
LGALS2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 152 amino acids (1-132 a.a.) and having a molecular mass of 16.8 kDa. The LGALS2 is fused to 20 amino acid His-Tag at N-terminus and pur 1 * 5 µG
Catalog # BIRBORB80329-5
Supplier:  Biorbyt