ContactUsLinkComponent
LGALS1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids and having a molecular mass of 14.5 kDa.The LGALS1 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80328-10
Supplier:  Biorbyt
LGALS1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids and having a molecular mass of 14.5 kDa.The LGALS1 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80328-10
Supplier:  Biorbyt