ContactUsLinkComponent
Thymosin b4 is 43 amino acid peptide which is regarded as the main intracellular G-actin sequestering peptide.It has molecular weigh t of 4963.55 Da, and its molecular formula is: C212H350N56O78S1. Extracellular Thymosin b4 may contribute to 1 * 5 mg
Catalog # BIRBORB80762-5
Supplier:  Biorbyt
Thymosin b4 is 43 amino acid peptide which is regarded as the main intracellular G-actin sequestering peptide.It has molecular weigh t of 4963.55 Da, and its molecular formula is: C212H350N56O78S1. Extracellular Thymosin b4 may contribute to 1 * 5 mg
Catalog # BIRBORB80762-5
Supplier:  Biorbyt