ContactUsLinkComponent
Terlipressin contains 12 amino acids Gly-Gly-Gly-c[Cys-Tyr-Phe-Gln-Asn-Cys]-Pro-Lys-Gly-NH2 and having molecular weight of 1227.4 Da lton. 1 * 250 mg
Catalog # BIRBORB80760-250
Dodavatel:  Biorbyt
Terlipressin contains 12 amino acids Gly-Gly-Gly-c[Cys-Tyr-Phe-Gln-Asn-Cys]-Pro-Lys-Gly-NH2 and having molecular weight of 1227.4 Da lton. 1 * 250 mg
Catalog # BIRBORB80760-250
Dodavatel:  Biorbyt
Související informace
Frequently Bought Together