ContactUsLinkComponent
Rat FGF9 Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 205 amino acids and having a mol ecular mass of 23308 Dalton.The FGF-9 Mouse Recombinant is purified by proprietary chromatographic technique 1 * 10 µG
Catalog # BIRBORB80315-10
Supplier:  Biorbyt
Rat FGF9 Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 205 amino acids and having a mol ecular mass of 23308 Dalton.The FGF-9 Mouse Recombinant is purified by proprietary chromatographic technique 1 * 10 µG
Catalog # BIRBORB80315-10
Supplier:  Biorbyt