ContactUsLinkComponent
Procalcitonin Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 114 amino acids fragm ent (3-116) having a molecular mass of 17.13 kDa and an amino-terminal hexahistidine tag. The PCT is purifie 1 * 10 µG
  BIRBORB80751-10
 :  Biorbyt
Procalcitonin Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 114 amino acids fragm ent (3-116) having a molecular mass of 17.13 kDa and an amino-terminal hexahistidine tag. The PCT is purifie 1 * 10 µG
  BIRBORB80751-10
 :  Biorbyt
Frequently Bought Together