ContactUsLinkComponent
PMSG is complex glycoprotein obtained from the serum of pregnant mares. This 43-63 kda protein is capable of supplementing and being substituted for the follicle stimulating gonadotrophin and interstitial cell-stimulating hormone of the ant 1 * 1 items
Catalog # BIRBORB80748-1000
Supplier:  Biorbyt
PMSG is complex glycoprotein obtained from the serum of pregnant mares. This 43-63 kda protein is capable of supplementing and being substituted for the follicle stimulating gonadotrophin and interstitial cell-stimulating hormone of the ant 1 * 1 items
Catalog # BIRBORB80748-1000
Supplier:  Biorbyt