ContactUsLinkComponent
FGF acidic Bovine (FGF-1) purified from Bovine Brain contains 17 kDa and 20 kDa polypeptide chain. The 17 kDa peptide is derived fro m the 20K peptide by restricted proteolysis. (See Jaye et all). The FGF acidic is purified by proprietary ch 1 * 1 µG
Catalog # BIRBORB80308-1
Supplier:  Biorbyt
FGF acidic Bovine (FGF-1) purified from Bovine Brain contains 17 kDa and 20 kDa polypeptide chain. The 17 kDa peptide is derived fro m the 20K peptide by restricted proteolysis. (See Jaye et all). The FGF acidic is purified by proprietary ch 1 * 1 µG
Catalog # BIRBORB80308-1
Supplier:  Biorbyt