ContactUsLinkComponent
CD105 Mouse Recombinant extracellular domain produced in baculovirus is homodimeric, glycosylated, Polypeptide containing 581 amino acids and having a molecular mass of 61 kDa but as result of glycosylation, migrates at 75-85 kDa under redu 1 * 10 µG
Catalog # BIRBORB80305-10
Supplier:  Biorbyt
CD105 Mouse Recombinant extracellular domain produced in baculovirus is homodimeric, glycosylated, Polypeptide containing 581 amino acids and having a molecular mass of 61 kDa but as result of glycosylation, migrates at 75-85 kDa under redu 1 * 10 µG
Catalog # BIRBORB80305-10
Supplier:  Biorbyt