EBI Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 209 amino acids fragment (21-22 9) having molecular weight of 34kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The EBI is purif 1 * 10 µG
Catalog # BIRBORB80296-10
Supplier: Biorbyt
Frequently Bought Together