ContactUsLinkComponent
The molecular formula is C50H69N15O9. It has the amino acid sequence of Ac-Nle-cyclo [Asp-His-D-Phe-Arg-Trp-Lys]-NH2 and molecular w eight of 1024.2 Dalton. The CAS Number is 121062-08-6. 1 * 20 mg
Catalog # BIRBORB80743-20
Supplier:  Biorbyt
The molecular formula is C50H69N15O9. It has the amino acid sequence of Ac-Nle-cyclo [Asp-His-D-Phe-Arg-Trp-Lys]-NH2 and molecular w eight of 1024.2 Dalton. The CAS Number is 121062-08-6. 1 * 20 mg
Catalog # BIRBORB80743-20
Supplier:  Biorbyt