ContactUsLinkComponent
EBI3 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 209 amino acids and having a mo lecular mass of 23.3 kDa. The Human EBI3 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80295-10
Supplier:  Biorbyt
EBI3 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 209 amino acids and having a mo lecular mass of 23.3 kDa. The Human EBI3 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80295-10
Supplier:  Biorbyt