ContactUsLinkComponent
Lutenizing Hormone Releasing Hormone Human Synthetic is a single, non-glycosylated, polypeptide chain containing 10 amino acids and having a molecular mass of 1182..3 Dalton. The molecular formula is C55H75N17O13. The CAS Number is 71447-49 1 * 100 mg
Catalog # BIRBORB80739-100
Supplier:  Biorbyt
Lutenizing Hormone Releasing Hormone Human Synthetic is a single, non-glycosylated, polypeptide chain containing 10 amino acids and having a molecular mass of 1182..3 Dalton. The molecular formula is C55H75N17O13. The CAS Number is 71447-49 1 * 100 mg
Catalog # BIRBORB80739-100
Supplier:  Biorbyt