ContactUsLinkComponent
Inhibin-Alpha chain Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids fragment (233-366) and having an amino-terminal hexahistidine tag, having total molecular weight of 19.2 kD 1 * 2 µG
Catalog # BIRBORB80735-2
Supplier:  Biorbyt
Inhibin-Alpha chain Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids fragment (233-366) and having an amino-terminal hexahistidine tag, having total molecular weight of 19.2 kD 1 * 2 µG
Catalog # BIRBORB80735-2
Supplier:  Biorbyt