ContactUsLinkComponent
Inhibin-Alpha chain Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids fragment (233-366) and having an amino-terminal hexahistidine tag, having total molecular weight of 19.2 kD 1 * 10 µG
  BIRBORB80735-10
 :  Biorbyt
Inhibin-Alpha chain Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 134 amino acids fragment (233-366) and having an amino-terminal hexahistidine tag, having total molecular weight of 19.2 kD 1 * 10 µG
  BIRBORB80735-10
 :  Biorbyt
Frequently Bought Together