ContactUsLinkComponent
Mouse TNFSF11/RANKL ELISA Kit. Receptor activator of nuclear factor kappa-B ligand(RANKL), also known as tumor necrosis factor ligan d superfamily member 11 (TNFSF11), is a protein that in humans is encoded by the TNFSF11 gene. This gene enc 1 * 96 items
Catalog # BIRBORB138069-96
Supplier:  Biorbyt
Mouse TNFSF11/RANKL ELISA Kit. Receptor activator of nuclear factor kappa-B ligand(RANKL), also known as tumor necrosis factor ligan d superfamily member 11 (TNFSF11), is a protein that in humans is encoded by the TNFSF11 gene. This gene enc 1 * 96 items
Catalog # BIRBORB138069-96
Supplier:  Biorbyt