ContactUsLinkComponent
Human FGF-13 is encoded by the FGF13 gene. FGF-13 belongs to the FGF-11 subfamily which has four members FGF-11 toFGF-14. These four members were initially referred to as fibroblast growth factor homologous factors (FHFs) because they haveh 1 * 20 µG
Catalog # BIRBORB107718-20
Supplier:  Biorbyt
Human FGF-13 is encoded by the FGF13 gene. FGF-13 belongs to the FGF-11 subfamily which has four members FGF-11 toFGF-14. These four members were initially referred to as fibroblast growth factor homologous factors (FHFs) because they haveh 1 * 20 µG
Catalog # BIRBORB107718-20
Supplier:  Biorbyt