ContactUsLinkComponent
FGF-10 was originally identified from rat embryos by homology-based polymerase chain reaction. Rat FGF-10 sharesapproximately 95% am ino acid sequence identity with human FGF-10. Among the FGF family members, FGF-10 is most closelyrelated to 1 * 5 µG
Catalog # BIRBORB107717-5
Supplier:  Biorbyt
FGF-10 was originally identified from rat embryos by homology-based polymerase chain reaction. Rat FGF-10 sharesapproximately 95% am ino acid sequence identity with human FGF-10. Among the FGF family members, FGF-10 is most closelyrelated to 1 * 5 µG
Catalog # BIRBORB107717-5
Supplier:  Biorbyt