ContactUsLinkComponent
Fibroblast growth factor 9 (FGF-9) belongs to the large FGF family which has at least 23 members. All FGF family members areheparin binding growth factors with a core 120 amino acid (a.a.) FGF domain that allows for a common tertiary struct 1 * 20 µG
Catalog # BIRBORB107715-20
Supplier:  Biorbyt
Fibroblast growth factor 9 (FGF-9) belongs to the large FGF family which has at least 23 members. All FGF family members areheparin binding growth factors with a core 120 amino acid (a.a.) FGF domain that allows for a common tertiary struct 1 * 20 µG
Catalog # BIRBORB107715-20
Supplier:  Biorbyt