ContactUsLinkComponent
Epithelial cell-derived neutrophil-activating peptide 78 (ENA-78) is a member of the CXC subfamily of chemokines that has theGlu-Leu -Arg (ELR) motif preceding the CXC motif. Similar to other ELR containing CXC chemokines, ENA-78 is a potent 1 * 20 µG
Catalog # BIRBORB107705-20
Supplier:  Biorbyt
Epithelial cell-derived neutrophil-activating peptide 78 (ENA-78) is a member of the CXC subfamily of chemokines that has theGlu-Leu -Arg (ELR) motif preceding the CXC motif. Similar to other ELR containing CXC chemokines, ENA-78 is a potent 1 * 20 µG
Catalog # BIRBORB107705-20
Supplier:  Biorbyt