ContactUsLinkComponent
Chemokine (C-C motif) ligand 6 (CCL6) is a small cytokine belonging to the CC chemokine family that has only been identifiedin roden ts. Rat CCL6 is encoded by Ccl6 gene and the completed sequence is 115 amino acids (a.a.) containing 21 a.a. 1 * 2 µG
Catalog # BIRBORB107693-2
Supplier:  Biorbyt
Chemokine (C-C motif) ligand 6 (CCL6) is a small cytokine belonging to the CC chemokine family that has only been identifiedin roden ts. Rat CCL6 is encoded by Ccl6 gene and the completed sequence is 115 amino acids (a.a.) containing 21 a.a. 1 * 2 µG
Catalog # BIRBORB107693-2
Supplier:  Biorbyt