ContactUsLinkComponent
IL-22 is belonging to IL-10 family of regulatory cytokines which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Membersof thi s family share partial homology in their amino acid sequences, but they are dissimilar in their biological f 1 * 2 µG
Catalog # BIRBORB107760-2
Supplier:  Biorbyt
IL-22 is belonging to IL-10 family of regulatory cytokines which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Membersof thi s family share partial homology in their amino acid sequences, but they are dissimilar in their biological f 1 * 2 µG
Catalog # BIRBORB107760-2
Supplier:  Biorbyt