ContactUsLinkComponent
Interleukin-7 (IL-7) is encoded by the IL7 gene and secreted by stromal cells in the red marrow and thymus. It binds to the IL-7rece ptor, a heterodimer consisting of IL-7 receptor alpha and IL-2 receptor gamma chain. IL-7 stimulates the dif 1 * 10 µG
Catalog # BIRBORB107751-10
Supplier:  Biorbyt
Interleukin-7 (IL-7) is encoded by the IL7 gene and secreted by stromal cells in the red marrow and thymus. It binds to the IL-7rece ptor, a heterodimer consisting of IL-7 receptor alpha and IL-2 receptor gamma chain. IL-7 stimulates the dif 1 * 10 µG
Catalog # BIRBORB107751-10
Supplier:  Biorbyt