ContactUsLinkComponent
Human CCL14 is belonging to the CC chemokine family. It is encoded by the gene CCL14. CCL14 has two isoforms, CCL14a (HCC-1) and CCL 14b (HCC-3). The sequence of HCC-3 differs from HCC-1 as follow: 27-27 R-QTGGKPKVVKIQLKLVG. CCL14 was first 1 * 2 µG
Catalog # BIRBORB107735-2
Supplier:  Biorbyt
Human CCL14 is belonging to the CC chemokine family. It is encoded by the gene CCL14. CCL14 has two isoforms, CCL14a (HCC-1) and CCL 14b (HCC-3). The sequence of HCC-3 differs from HCC-1 as follow: 27-27 R-QTGGKPKVVKIQLKLVG. CCL14 was first 1 * 2 µG
Catalog # BIRBORB107735-2
Supplier:  Biorbyt