ContactUsLinkComponent
Fibroblast growth factor-23 (FGF-23) belongs to the large FGF family which has at least 23 members. All FGF family membersare hepari n binding growth factors with a core 120 amino acid (a.a.) FGF domain that allows for a common tertiary stru 1 * 20 µG
Catalog # BIRBORB107724-20
Supplier:  Biorbyt
Fibroblast growth factor-23 (FGF-23) belongs to the large FGF family which has at least 23 members. All FGF family membersare hepari n binding growth factors with a core 120 amino acid (a.a.) FGF domain that allows for a common tertiary stru 1 * 20 µG
Catalog # BIRBORB107724-20
Supplier:  Biorbyt