ContactUsLinkComponent
Fibroblast growth factor 21 (FGF-21) encoded by the FGF-21 gene belongs to the large FGF family and it is specifically inducedby HMG CS2 activity. In mice, brown adipose tissue becomes a source of systemic FGF-21 after cold exposure. FGF-21s 1 * 25 µG
Catalog # BIRBORB107723-25
Supplier:  Biorbyt
Fibroblast growth factor 21 (FGF-21) encoded by the FGF-21 gene belongs to the large FGF family and it is specifically inducedby HMG CS2 activity. In mice, brown adipose tissue becomes a source of systemic FGF-21 after cold exposure. FGF-21s 1 * 25 µG
Catalog # BIRBORB107723-25
Supplier:  Biorbyt