ContactUsLinkComponent
Micro-Centrifugal Filter 750µL, 0.20µm Nylon Membrane, Nonsterile 1 * 100 items
Catalog # CRMAF2517-3
Supplier:  CHROMACOL
Micro-Centrifugal Filter 750µL, 0.20µm Nylon Membrane, Nonsterile 1 * 100 items
Catalog # CRMAF2517-3
Supplier:  CHROMACOL
Frequently Bought Together