Multi-parameter testers, Pocket Pro™+
Tester for pH, conductivity, TDS, salinity and temperature, Pocket Pro™+ Multi 2
Catalog # 665-0400
Supplier: Hach
Jump to:
Product Details & Documents
Convenient portable testers with replaceable sensors for measuring conductivity, TDS, salinity, temperature and pH values (only Multi 2). Backed up with built in performance diagnostics, users never have to guess when to clean or calibrate the sensor.
- With pH glass sensing element
- Easy calibration steps and built-in diagnostics for testers
- Easy-to-replace AAA batteries and a convenient sample cup make maintenance and measurements simple
- Large LCD screen is easy to read and backlight allows for taking measurements in almost any environment
- Intuitive user interface
- Battery life: 450 hours (200 hours with backlight on)
- One year warranty on meter and sensor
Ideal for field measurements in beverages, industrial water, wasterwater and swimming pool water.
: Testers are supplied with 4 AAA batteries, lanyard, and quick start guide. Calibration solutions and other accessories are ordered separately. Please contact your Avantor representative for more information.
Specifications
- Description:Tester for pH, conductivity, TDS, salinity and temperature, Pocket Pro™+ Multi 2
- Seq:03
- Compliance:IP 67
- pH range:0,00 - 14,00
- Conductivity resolution:0,1 µS/cm (0,0 - 199,9 µS/cm), 1 µS/cm (200 - 1999 µS/cm), 0,01 mS/cm (2,00 - 20,00 mS/cm)
- Power supply:4 × AAA batteries
- pH calibration:3 points (automatic), 2 points (custom)
- Temperature resolution:0,1 °C
- TDS resolution:0,1 ppm (0,0 - 99,9 ppm), 1 ppm (100 - 999 ppm), 0,01 ppt (1,00 - 10,00 ppt)
- Temperature accuracy:±0,5 °C
- Salinity accuracy:±1%
- Salinity resolution:0,1 ppm (0,0 - 99,9 ppm), 1 ppm (100 - 999 ppm), 0,01 ppt (1,0 - 10,00 ppt), 0,01% (0,0 - 1,00%)
- TDS accuracy:±1%
- Weight:0,135 kg (with batteries)
- Cat. No.:665-0400
- Conductivity accuracy:±1%
- Conductivity calibration:3 points (automatic), 1 point (custom)
- pH accuracy:±0,02
- pH resolution:0,01
- Model:Pocket Pro™+ Multi 2
- Conductivity range:Auto-ranging: 0,0 - 199,9 µS/cm / 200 - 1999 µS/cm / 2,00 - 19,99 mS/cm
- TDS range:Auto-ranging: 0,0 - 99,9 ppm / 100 - 999 ppm / 1,00 - 10,00 ppt
- W×D×H:37×30×170 mm
- Temperature range:0,0...+50 °C
- Salinity range:Auto-ranging: 0,00 - 99,9 ppm / 100 - 999 ppm / 1,0 - 10,00 ppt / 0,00 - 1,00%
- Display:LCD with backlight
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement sensor for conductivity/TDS/salinity/temperature measurements, for Multi 1 meterReplacement sensor for conductivity/TDS/salinity/temperature measurements, for Multi 1 meterHACH9532701Log in for availability1 * 1 KS - 4 650,00 Kč
Product Information
- Catalog #pH valuePack typeAvailabilityPrice
- pH buffer solution combination pack, single-usepH buffer solution combination pack, single-useHACH27699204,01/7,00Foil pouchLog in for availability1 * 1 SET - 1 580,00 Kč
Product Information
- Catalog #AvailabilityPrice
- Replacement sensor for pH/conductivity/TDS/salinity/temperature measurements, for Multi 2 meterReplacement sensor for pH/conductivity/TDS/salinity/temperature measurements, for Multi 2 meterHACH9532801Log in for availability1 * 1 KS - 5 370,00 Kč



