ContactUsLinkComponent
Uv convenience kit. includes 2 x open-top uv quartz cell, 1mm, 3.ml vol. cell tray with capacity for 16 cells lens cleaning paper (lint free) and multi-function tool. 1 * 1 items
Catalog # HEWL5067-4666
Supplier:  Agilent
Uv convenience kit. includes 2 x open-top uv quartz cell, 1mm, 3.ml vol. cell tray with capacity for 16 cells lens cleaning paper (lint free) and multi-function tool. 1 * 1 items
Catalog # HEWL5067-4666
Supplier:  Agilent
Frequently Bought Together