ContactUsLinkComponent
Db-vrx, 2 m, .18 mm, 1. um. ltm column toroid assembly for agilent 5975t. replacement column for customer's existing column module h ardware. 1 * 1 items
Catalog # HEWL221-1524LTM
Supplier:  Agilent
Db-vrx, 2 m, .18 mm, 1. um. ltm column toroid assembly for agilent 5975t. replacement column for customer's existing column module h ardware. 1 * 1 items
Catalog # HEWL221-1524LTM
Supplier:  Agilent
Frequently Bought Together