ContactUsLinkComponent
Concentric nebulizer for mp-aes. used with 41/42 series. recommended for use with aqueous solutions and normal digests 1 * 1 items
Catalog # HEWLG8000-70004
Supplier:  Agilent
undefined
Concentric nebulizer for mp-aes. used with 41/42 series. recommended for use with aqueous solutions and normal digests 1 * 1 items
Catalog # HEWLG8000-70004
Supplier:  Agilent
Frequently Bought Together