ContactUsLinkComponent
Crosslab piston seal/support ring kit, reverse phase, contains 2 piston seals and 1 support ring, for ultimate 3 iso-31a, lpg-34a/m, dgp-36a/m, and hpg-3xa/m pumps, tmo 1 * 1 items
Catalog # HEWL8002-0923
Supplier:  Agilent
Crosslab piston seal/support ring kit, reverse phase, contains 2 piston seals and 1 support ring, for ultimate 3 iso-31a, lpg-34a/m, dgp-36a/m, and hpg-3xa/m pumps, tmo 1 * 1 items
Catalog # HEWL8002-0923
Supplier:  Agilent
Frequently Bought Together