ContactUsLinkComponent
Rear seal wash tubing kit u3 pump tmo crosslab tubing kit, rear seal wash system, contains 1 silicone tubing (2.8mm od x 1.3mm id x 1.5m l), 1 pharmed tubing for peristaltic pump (3.2mm od x 1.6mm id x 18mm l), 7 tube connectors for 1.-2. mm id tubin 1 * 1 items
Catalog # HEWL8002-0916
Supplier:  Agilent
undefined
Rear seal wash tubing kit u3 pump tmo crosslab tubing kit, rear seal wash system, contains 1 silicone tubing (2.8mm od x 1.3mm id x 1.5m l), 1 pharmed tubing for peristaltic pump (3.2mm od x 1.6mm id x 18mm l), 7 tube connectors for 1.-2. mm id tubin 1 * 1 items
Catalog # HEWL8002-0916
Supplier:  Agilent
Frequently Bought Together