ContactUsLinkComponent
Bio-inert standard flow cell, rfid tag 1 mm, 13 ul, 12 bar, for g1315c/d and g1365c/d 1 * 1 items
Catalog # HEWLG5615-60022
Supplier:  Agilent
Bio-inert standard flow cell, rfid tag 1 mm, 13 ul, 12 bar, for g1315c/d and g1365c/d 1 * 1 items
Catalog # HEWLG5615-60022
Supplier:  Agilent
Frequently Bought Together