ContactUsLinkComponent
Kit gta 12 zeeman gfaa supplies 1/pk incl.2 sets graphite electrodes shroud  5 pks omega platform tubes  1ul syringe 1 pk capillary assemblies  1 pk 1ml beakers and 2 pks sample vials 1 * 1 items
Catalog # VARI190068000
Supplier:  VARIAN
Kit gta 12 zeeman gfaa supplies 1/pk incl.2 sets graphite electrodes shroud  5 pks omega platform tubes  1ul syringe 1 pk capillary assemblies  1 pk 1ml beakers and 2 pks sample vials 1 * 1 items
Catalog # VARI190068000
Supplier:  VARIAN
Frequently Bought Together