ContactUsLinkComponent
Sax polymer - box, 1x 1ml tubes,3 mg sampliq solid phase extraction (spe) 1 * 100 items
Catalog # HEWL5982-3313
Supplier:  Agilent
Sax polymer - box, 1x 1ml tubes,3 mg sampliq solid phase extraction (spe) 1 * 100 items
Catalog # HEWL5982-3313
Supplier:  Agilent
Frequently Bought Together