ContactUsLinkComponent
Comprehensive spares kit for agilent icp-ms systems fitted with x-lens. excl. interface cones - select required option to add. suits 77x/e and 8 series icp-ms 1 * 1 items
Catalog # HEWLG3690A
Supplier:  Agilent
Comprehensive spares kit for agilent icp-ms systems fitted with x-lens. excl. interface cones - select required option to add. suits 77x/e and 8 series icp-ms 1 * 1 items
Catalog # HEWLG3690A
Supplier:  Agilent
Frequently Bought Together