ContactUsLinkComponent
Digestion tubes polypropylene, 5 ml, 5/pk. 3mm od x 115mm. features conical base, wide neck + threaded top for screw cap. metal free polyethylene liner. graduated to 5 ml at 5 ml intervals 1 * 500 items
Catalog # VARI190047900
Supplier:  VARIAN
Digestion tubes polypropylene, 5 ml, 5/pk. 3mm od x 115mm. features conical base, wide neck + threaded top for screw cap. metal free polyethylene liner. graduated to 5 ml at 5 ml intervals 1 * 500 items
Catalog # VARI190047900
Supplier:  VARIAN
Frequently Bought Together